Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lztfl1
Entrez Gene ID
316102
Gene Name
leucine zipper transcription factor-like 1
Gene Aliases
-
Chromosome Location
Chr.8: 132774393 - 132790922 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024266.1 NP_001019437.1 6 - 7 568 129
XM_006244191.1 XP_006244253.1 6 - 7 712 129
GenBank mRNA BC078913.1 - 6 - 7 664 129
BC092585.1 - 6 - 7 568 129

View Full Product Details