Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dgcr2
Entrez Gene ID
360742
Gene Name
DiGeorge syndrome critical region gene 2
Gene Aliases
-
Chromosome Location
Chr.11: 87242441 - 87292914 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012146.1 NP_001012146.1 9 - 10 1663 59
XM_008768890.1 XP_008767112.1 9 - 10 1646 59
GenBank mRNA BC081905.1 - 9 - 10 1663 59
FQ211720.1 - 9 - 10 1587 59
FQ228410.1 - 9 - 10 1565 59

Gene Symbol
LOC100910275
Entrez Gene ID
100910275
Gene Name
integral membrane protein DGCR2/IDD-like
Gene Aliases
-
Chromosome Location
Chr.6: 138351215 - 138401703 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003750241.2 XP_003750289.1 9 - 10 1662 59
XM_006240675.2 XP_006240737.1 9 - 10 1639 59

Gene Symbol
LOC103693608
Entrez Gene ID
103693608
Gene Name
integral membrane protein DGCR2/IDD-like
Gene Aliases
-
Chromosome Location
Chr.11: 87765024 - 87775583 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768936.1 XP_008767158.1 3 - 4 786 59

View Full Product Details