The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105864.1 | NP_001099334.1 | 9 - 10 | 1613 | 86 |
XM_006248688.2 | XP_006248750.1 | 9 - 10 | 1616 | 86 |