Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cldn16
Entrez Gene ID
155268
Gene Name
claudin 16
Gene Aliases
Pcln1
Chromosome Location
Chr.11: 77683942 - 77703232 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131905.1 NP_571980.1 2 - 3 543 76
GenBank mRNA AF333099.1 - 2 - 3 543 76

View Full Product Details