The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198133.2 | NP_937766.1 | 4 - 5 | 397 | 90 |
GenBank mRNA | AY321314.1 | - | 4 - 5 | 397 | 90 |