The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008768878.1 | XP_008767100.1 | 2 - 3 | 290 | 77 |
GenBank mRNA | FQ211899.1 | - | 2 - 3 | 109 | 77 |