The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053933.1 | NP_446385.1 | 1 - 2 | 2388 | 64 |
GenBank mRNA | AB045585.1 | - | 1 - 2 | 890 | 64 |
AB113387.1 | - | 1 - 2 | 2388 | 64 | |
AY573978.1 | - | 1 - 2 | 2388 | 64 |