Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stk32a
Entrez Gene ID
364858
Gene Name
serine/threonine kinase 32A
Gene Aliases
-
Chromosome Location
Chr.18: 37597305 - 37706884 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191894.1 NP_001178823.1 7 - 8 809 70

View Full Product Details