Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pfdn1
Entrez Gene ID
361310
Gene Name
prefoldin subunit 1
Gene Aliases
-
Chromosome Location
Chr.18: 29233987 - 29290480 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108427.1 NP_001101897.1 2 - 3 220 81
XM_006254598.1 XP_006254660.1 2 - 3 253 81

View Full Product Details