Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ik
Entrez Gene ID
291659
Gene Name
IK cytokine
Gene Aliases
CSA2, RED
Chromosome Location
Chr.18: 29587901 - 29602705 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005537.1 NP_001005537.1 18 - 19 1734 66
GenBank mRNA BC081870.1 - 18 - 19 1734 66
FQ212525.1 - 18 - 19 1686 66
FQ214775.1 - 15 - 16 1444 66

View Full Product Details