Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wnt8a
Entrez Gene ID
291678
Gene Name
wingless-type MMTV integration site family, member 8A
Gene Aliases
-
Chromosome Location
Chr.18: 27331236 - 27336829 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106155.1 NP_001099625.1 2 - 3 150 62

View Full Product Details