Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ecscr
Entrez Gene ID
689248
Gene Name
endothelial cell surface expressed chemotaxis and apoptosis regulator
Gene Aliases
Aria, Ecscr-ps1
Chromosome Location
Chr.18: 28507167 - 28516237 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772057.1 XP_008770279.1 3 - 4 311 59

View Full Product Details