The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199396.1 | NP_955428.1 | 7 - 8 | 2265 | 69 |
XM_006242213.2 | XP_006242275.1 | 6 - 7 | 1698 | 69 | |
XM_006242214.2 | XP_006242276.1 | 7 - 8 | 4162 | 69 | |
XM_006242215.2 | XP_006242277.1 | 7 - 8 | 2093 | 69 | |
GenBank mRNA | AF399843.1 | - | 4 - 5 | 1454 | 69 |