Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpf2
Entrez Gene ID
294436
Gene Name
ribosome production factor 2 homolog (S. cerevisiae)
Gene Aliases
Bxdc1
Chromosome Location
Chr.20: 45321381 - 45342865 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106391.2 NP_001099861.1 3 - 4 215 113
GenBank mRNA BC158719.1 - 3 - 4 215 113

View Full Product Details