Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Creb3l3
Entrez Gene ID
314638
Gene Name
cAMP responsive element binding protein 3-like 3
Gene Aliases
-
Chromosome Location
Chr.7: 11490852 - 11499287 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012115.1 NP_001012115.1 4 - 5 643 126
GenBank mRNA BC089877.1 - 4 - 5 643 126

View Full Product Details