Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hmg20b
Entrez Gene ID
362825
Gene Name
high mobility group 20 B
Gene Aliases
-
Chromosome Location
Chr.7: 11238594 - 11243312 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108731.1 NP_001102201.1 8 - 9 1249 70
XM_006240944.2 XP_006241006.1 9 - 10 1146 70
XM_006240946.2 XP_006241008.1 9 - 10 1068 70
GenBank mRNA FQ220489.1 - 4 - 5 454 70

View Full Product Details