Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pias4
Entrez Gene ID
362827
Gene Name
protein inhibitor of activated STAT, 4
Gene Aliases
-
Chromosome Location
Chr.7: 11414565 - 11428090 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100757.1 NP_001094227.1 7 - 8 937 82
GenBank mRNA BC169045.1 - 7 - 8 937 82

View Full Product Details