Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp4f37
Entrez Gene ID
691312
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 37
Gene Aliases
-
Chromosome Location
Chr.7: 14589520 - 14614869 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271352.1 NP_001258281.1 4 - 5 736 84

Gene Symbol
Cyp4f5
Entrez Gene ID
286905
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 5
Gene Aliases
-
Chromosome Location
Chr.7: 14559806 - 14574345 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173124.1 NP_775147.1 - 416 84
XM_006241051.2 XP_006241113.1 3 - 4 485 84
XM_008765158.1 XP_008763380.1 3 - 4 487 84
GenBank mRNA BC129085.1 - 3 - 4 377 84
U39207.1 - - 416 84

View Full Product Details