Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plekhj1
Entrez Gene ID
314634
Gene Name
pleckstrin homology domain containing, family J member 1
Gene Aliases
-
Chromosome Location
Chr.7: 11784314 - 11786476 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240929.1 XP_006240991.1 4 - 5 388 65
XM_006240930.1 XP_006240992.1 4 - 5 388 65

View Full Product Details