The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039003.1 | NP_001034092.1 | 12 - 13 | 1453 | 112 |
XM_006227813.2 | XP_006227875.1 | 14 - 15 | 2082 | 112 | |
XM_006227814.1 | XP_006227876.1 | 12 - 13 | 1746 | 112 | |
XM_008758680.1 | XP_008756902.1 | 11 - 12 | 2156 | 112 | |
GenBank mRNA | BC100107.1 | - | 12 - 13 | 1453 | 112 |