Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galntl5
Entrez Gene ID
499968
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 5
Gene Aliases
Galnt15
Chromosome Location
Chr.4: 6333049 - 6450151 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025148.1 NP_001020319.1 5 - 6 736 140
XM_006235958.2 XP_006236020.1 4 - 5 699 140
GenBank mRNA BC078995.1 - 5 - 6 736 140

View Full Product Details