Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galntl5
Entrez Gene ID
499968
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 5
Gene Aliases
Galnt15
Chromosome Location
Chr.4: 6333049 - 6450151 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025148.1 NP_001020319.1 7 - 8 1102 106
XM_006235958.2 XP_006236020.1 6 - 7 1065 106
GenBank mRNA BC078995.1 - 7 - 8 1102 106

View Full Product Details