The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025148.1 | NP_001020319.1 | 8 - 9 | 1252 | 106 |
XM_006235958.2 | XP_006236020.1 | 7 - 8 | 1215 | 106 | |
GenBank mRNA | BC078995.1 | - | 8 - 9 | 1252 | 106 |