Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Elovl3
Entrez Gene ID
309449
Gene Name
ELOVL fatty acid elongase 3
Gene Aliases
-
Chromosome Location
Chr.1: 265883353 - 265887181 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107602.1 NP_001101072.1 3 - 4 556 67

View Full Product Details