The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105973.1 | NP_001099443.1 | 1 - 2 | 119 | 65 |
XM_006250284.2 | XP_006250346.1 | 1 - 2 | 1532 | 65 |