Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slamf8
Entrez Gene ID
289237
Gene Name
SLAM family member 8
Gene Aliases
-
Chromosome Location
Chr.13: 90967862 - 90978756 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105973.1 NP_001099443.1 4 - 5 863 87

View Full Product Details