Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hhat
Entrez Gene ID
289344
Gene Name
hedgehog acyltransferase
Gene Aliases
RGD1311746
Chromosome Location
Chr.13: 111235325 - 111489067 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769894.1 XP_008768116.1 10 - 11 2132 119

View Full Product Details