The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019312.2 | NP_062185.2 | 6 - 7 | 3004 | 90 |
GenBank mRNA | AJ242781.1 | - | - | 2553 | 90 |
X74227.1 | - | - | 2327 | 90 |