Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pvrl4
Entrez Gene ID
498281
Gene Name
poliovirus receptor-related 4
Gene Aliases
RGD1559826
Chromosome Location
Chr.13: 89755665 - 89774185 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109076.1 NP_001102546.1 6 - 7 1622 61
XM_006250256.2 XP_006250318.1 6 - 7 1591 61
GenBank mRNA FQ228572.1 - 5 - 6 1142 61
FQ228642.1 - 5 - 6 1142 61

View Full Product Details