Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nipal1
Entrez Gene ID
289594
Gene Name
NIPA-like domain containing 1
Gene Aliases
Npal1, RGD1310092
Chromosome Location
Chr.14: 38104027 - 38128038 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106003.1 NP_001099473.1 1 - 2 129 152
XM_006250895.2 XP_006250957.1 1 - 2 143 152

View Full Product Details