The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031788.1 | NP_113976.1 | 2 - 3 | 984 | 84 |
XM_006250847.2 | XP_006250909.1 | - | 1063 | 84 | |
XM_006250848.1 | XP_006250910.1 | 3 - 4 | 1068 | 84 | |
XM_006250849.2 | XP_006250911.1 | 3 - 4 | 1326 | 84 | |
GenBank mRNA | AF037199.1 | - | 2 - 3 | 984 | 84 |