The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005882.1 | NP_001005882.1 | 4 - 5 | 1694 | 139 |
XM_006250949.2 | XP_006251011.1 | 5 - 6 | 2777 | 139 | |
XM_006250950.2 | XP_006251012.1 | 5 - 6 | 3675 | 139 | |
XM_006250951.2 | XP_006251013.1 | 7 - 8 | 2023 | 139 | |
XM_006250952.2 | XP_006251014.1 | 6 - 7 | 2249 | 139 | |
XM_006250954.2 | XP_006251016.1 | 6 - 7 | 2780 | 139 | |
XM_006250955.2 | XP_006251017.1 | 5 - 6 | 2968 | 139 | |
XM_008770152.1 | XP_008768374.1 | 5 - 6 | 2482 | 139 | |
GenBank mRNA | AY724525.1 | - | 5 - 6 | 1717 | 139 |
BC081995.1 | - | 4 - 5 | 1694 | 139 |