Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tecrl
Entrez Gene ID
364134
Gene Name
trans-2,3-enoyl-CoA reductase-like
Gene Aliases
Srd5a2l2
Chromosome Location
Chr.14: 26662851 - 26735878 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108860.1 NP_001102330.1 11 - 12 1153 91

View Full Product Details