The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106005.1 | NP_001099475.1 | 5 - 6 | 715 | 86 |
XM_006251036.2 | XP_006251098.1 | 5 - 6 | 759 | 86 | |
GenBank mRNA | BC168866.1 | - | 5 - 6 | 702 | 86 |