Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl15
Entrez Gene ID
309453
Gene Name
F-box and leucine-rich repeat protein 15
Gene Aliases
RGD1306444
Chromosome Location
Chr.1: 266075782 - 266078023 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107603.1 NP_001101073.1 2 - 3 502 50
GenBank mRNA FQ235302.1 - 2 - 3 494 50

View Full Product Details