Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alb
Entrez Gene ID
24186
Gene Name
albumin
Gene Aliases
Alb1, Albza
Chromosome Location
Chr.14: 19176275 - 19191793 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134326.2 NP_599153.2 7 - 8 860 65
GenBank mRNA BC085359.1 - 7 - 8 860 65
FQ209382.1 - 3 - 4 308 65
FQ209635.1 - 1 - 2 70 65
FQ209651.1 - 1 - 2 100 65
FQ209695.1 - 3 - 4 294 65
FQ209721.1 - 1 - 2 81 65
FQ209907.1 - 3 - 4 329 65
FQ209957.1 - 2 - 3 241 65
FQ210022.1 - 1 - 2 83 65
FQ210057.1 - 3 - 4 259 65
FQ210111.1 - 1 - 2 112 65
FQ210171.1 - 1 - 2 81 65
FQ210316.1 - 1 - 2 81 65
FQ210406.1 - 1 - 2 60 65
FQ210546.1 - 1 - 2 108 65
FQ210640.1 - 1 - 2 65 65
FQ210724.1 - 4 - 5 384 65
FQ210803.1 - 3 - 4 330 65
FQ210850.1 - 2 - 3 176 65
FQ210851.1 - 2 - 3 207 65
FQ210858.1 - 4 - 5 414 65
FQ210877.1 - 3 - 4 275 65
FQ210995.1 - 1 - 2 70 65
FQ211010.1 - 2 - 3 218 65
FQ211036.1 - 1 - 2 113 65
FQ211096.1 - 1 - 2 81 65
FQ211196.1 - 1 - 2 41 65
FQ211218.1 - 4 - 5 488 65
FQ211255.1 - 3 - 4 308 65
FQ211259.1 - 3 - 4 317 65
FQ211320.1 - 1 - 2 112 65
FQ211378.1 - 1 - 2 81 65
FQ218137.1 - 1 - 2 85 65
FQ218216.1 - 2 - 3 184 65
FQ218458.1 - 3 - 4 324 65
FQ218739.1 - 2 - 3 229 65
FQ218815.1 - 2 - 3 194 65
FQ219210.1 - 3 - 4 279 65
FQ219260.1 - 1 - 2 50 65
FQ219293.1 - 3 - 4 308 65
FQ219442.1 - 1 - 2 78 65
FQ219447.1 - 3 - 4 301 65
FQ219470.1 - 2 - 3 179 65
FQ219514.1 - 1 - 2 107 65
FQ219650.1 - 1 - 2 77 65
FQ219679.1 - 1 - 2 70 65
FQ219682.1 - 1 - 2 52 65

View Full Product Details