Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ambn
Entrez Gene ID
25376
Gene Name
ameloblastin
Gene Aliases
-
Chromosome Location
Chr.14: 21239887 - 21252534 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012900.1 NP_037032.1 4 - 5 280 99
GenBank mRNA U35097.1 - 4 - 5 280 99
Z50083.1 - 4 - 5 283 99

View Full Product Details