Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ambn
Entrez Gene ID
25376
Gene Name
ameloblastin
Gene Aliases
-
Chromosome Location
Chr.14: 21239887 - 21252534 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012900.1 NP_037032.1 9 - 10 781 130
GenBank mRNA U35097.1 - 9 - 10 781 130
Z50083.1 - 9 - 10 739 130
Z50084.1 - 5 - 6 448 130

View Full Product Details