Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rufy3
Entrez Gene ID
360921
Gene Name
RUN and FYVE domain containing 3
Gene Aliases
Ripx, Singar1
Chromosome Location
Chr.14: 21058575 - 21144743 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025127.1 NP_001020298.1 12 - 13 1687 105
XM_006250781.2 XP_006250843.1 12 - 13 2551 105
XM_006250782.2 XP_006250844.1 12 - 13 2760 105
XM_006250783.2 XP_006250845.1 12 - 13 2550 105
XM_006250784.2 XP_006250846.1 11 - 12 1276 105
XM_006250785.2 XP_006250847.1 11 - 12 1470 105
XM_008770044.1 XP_008768266.1 11 - 12 1488 105
XM_008770045.1 XP_008768267.1 11 - 12 1488 105
XM_008770046.1 XP_008768268.1 12 - 13 2745 105
XM_008770047.1 XP_008768269.1 11 - 12 1488 105
XM_008770048.1 XP_008768270.1 13 - 14 2604 105
GenBank mRNA BC089952.1 - 12 - 13 1687 105
EF538802.1 - 12 - 13 1292 105
EF577045.1 - 11 - 12 1238 105

View Full Product Details