Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Peli2
Entrez Gene ID
305835
Gene Name
pellino E3 ubiquitin protein ligase family member 2
Gene Aliases
RGD1565884
Chromosome Location
Chr.15: 24942218 - 25084250 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107259.1 NP_001100729.1 2 - 3 458 110

View Full Product Details