Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ero1l
Entrez Gene ID
171562
Gene Name
ERO1-like (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.15: 19619947 - 19655381 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138528.1 NP_612537.1 4 - 5 363 84
GenBank mRNA AF489855.1 - 4 - 5 350 84
AY071924.1 - 4 - 5 363 84
FQ228432.1 - - 208 84

View Full Product Details