Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ero1l
Entrez Gene ID
171562
Gene Name
ERO1-like (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.15: 19619947 - 19655381 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138528.1 NP_612537.1 5 - 6 439 127
GenBank mRNA AF489855.1 - 5 - 6 426 127
AY071924.1 - 5 - 6 439 127
FQ228432.1 - - 284 127

View Full Product Details