The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013185.1 | NP_001013203.1 | 14 - 15 | 1701 | 118 |
XM_006250387.1 | XP_006250449.1 | 14 - 15 | 1784 | 118 | |
GenBank mRNA | BC091388.1 | - | 14 - 15 | 1701 | 118 |