The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033666.1 | NP_001028838.1 | 2 - 3 | 430 | 89 |
GenBank mRNA | CF111187.1 | - | 2 - 3 | 390 | 89 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003751907.3 | XP_003751955.1 | 1 - 1 | 392 | 89 |