The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108595.1 | NP_001102065.1 | 4 - 5 | 398 | 104 |
XM_006235155.2 | XP_006235217.1 | 4 - 5 | 367 | 104 | |
GenBank mRNA | BC167020.1 | - | 4 - 5 | 358 | 104 |