Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppdpf
Entrez Gene ID
296470
Gene Name
pancreatic progenitor cell differentiation and proliferation factor
Gene Aliases
RGD1310660
Chromosome Location
Chr.3: 176690515 - 176692272 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006235746.2 XP_006235808.1 1 - 2 246 65

View Full Product Details