Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zyg11a
Entrez Gene ID
313482
Gene Name
zyg-11 family member A, cell cycle regulator
Gene Aliases
RGD1309582
Chromosome Location
Chr.5: 127790496 - 127828911 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_233348.8 XP_233348.6 12 - 13 2012 100

View Full Product Details