Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Skint4
Entrez Gene ID
313485
Gene Name
selection and upkeep of intraepithelial T cells 4
Gene Aliases
RGD1566327
Chromosome Location
Chr.5: 132005656 - 132043403 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238600.2 XP_006238662.2 4 - 5 830 105

View Full Product Details