Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zswim5
Entrez Gene ID
313524
Gene Name
zinc finger, SWIM-type containing 5
Gene Aliases
-
Chromosome Location
Chr.5: 135735825 - 135854525 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107963.1 NP_001101433.1 2 - 3 501 91
XM_008764009.1 XP_008762231.1 2 - 3 423 91

View Full Product Details