Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ube2q2
Entrez Gene ID
363065
Gene Name
ubiquitin-conjugating enzyme E2Q family member 2
Gene Aliases
RGD1307680
Chromosome Location
Chr.8: 59711223 - 59768423 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766429.1 XP_008764651.1 2 - 3 620 140

View Full Product Details